Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxy-6-methoxy-2-propylquinoline
4-Hydroxy-6-methoxy-2-propylquinoline
4-Hydroxy-6-methoxy-2-propylquinoline
APOSOR309305-1G
:
Apollo Scientific
:
4-Hydroxy-6-methoxy-2-propylquinoline
APOSOR309305-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
217.264 g/mol
:
Ambient
:
927800-85-9
:
C₁₃H₁₅NO₂
:
MFCD02684129