0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
8-Bromo-4-hydroxy-2-phenylquinoline
8-Bromo-4-hydroxy-2-phenylquinoline
8-Bromo-4-hydroxy-2-phenylquinoline
8-Bromo-4-hydroxy-2-phenylquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
8-Bromo-4-hydroxy-2-phenylquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
927800-73-5
:
Formula:
C₁₅H₁₀BrNO
MDL Number:
MFCD02684126
:
300.15 g/mol
Storage Temperature:
Ambient