Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxy-6-methyl-2-phenylquinoline
4-Hydroxy-6-methyl-2-phenylquinoline
4-Hydroxy-6-methyl-2-phenylquinoline
4-Hydroxy-6-methyl-2-phenylquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
4-Hydroxy-6-methyl-2-phenylquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
1148-49-8
:
Formula:
C₁₆H₁₃NO
MDL Number:
MFCD02179767
:
235.281 g/mol
Storage Temperature:
Ambient