Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxy-2-phenylquinoline
4-Hydroxy-2-phenylquinoline
4-Hydroxy-2-phenylquinoline
Corrosive
Irritant
4-Hydroxy-2-phenylquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
4-Hydroxy-2-phenylquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
1144-20-3
:
Formula:
C₁₅H₁₁NO
MDL Number:
MFCD00160565
:
221.254 g/mol
Melting Pt:
255…256 °C
:
GHS05
:
GHS07
Storage Temperature:
Ambient