Order Entry
Germany
ContactUsLinkComponent
2-Chloro-6-ethoxy-3-phenylquinoline
2-Chloro-6-ethoxy-3-phenylquinoline
Catalog # APOSOR309259-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-6-ethoxy-3-phenylquinoline
Catalog # APOSOR309259-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.752 g/mol
Storage Temperature:Ambient
CAS Number:1031928-18-3
Formula:C₁₇H₁₄ClNO
MDL Number:MFCD09998012