6-Ethoxy-4-hydrazino-2-methylquinoline hydrochloride
Catalog # APOSOR309213-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:253.728 g/mol
Storage Temperature:Ambient
CAS Number:1071572-37-6
Formula:C₁₂H₁₅N₃O·ClH
MDL Number:MFCD09787829