2-Amino-6-bromo-3-phenylquinoline hydrochloride
Catalog # APOSOR307892-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:335.626 g/mol
Storage Temperature:Ambient
CAS Number:1170960-09-4
Formula:C₁₅H₁₁BrN₂·ClH
MDL Number:MFCD10703381