Order Entry
Germany
ContactUsLinkComponent
 
3-(4-aminobenzylamino)-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSOR307776-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-aminobenzylamino)-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSOR307776-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :305.415 g/mol
Storage Temperature:Ambient
CAS Number:1189105-72-3
Formula:C₁₇H₂₇N₃O₂
MDL Number:MFCD12674814