N-(4-Aminobenzyl)-1-methylpiperidin-4-amine dihydrochloride
Catalog # APOSOR307772-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:292.248 g/mol
Storage Temperature:Ambient
CAS Number:1204810-58-1
Formula:C₁₃H₂₁N₃·2ClH
MDL Number:MFCD12674811