Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-tert-Butyl 3-phenylaziridine-2-carboxylate
cis-tert-Butyl 3-phenylaziridine-2-carboxylate
cis-tert-Butyl 3-phenylaziridine-2-carboxylate
cis-tert-Butyl 3-phenylaziridine-2-carboxylate
Supplier:
Apollo Scientific
Certificates
Expand image
cis-tert-Butyl 3-phenylaziridine-2-carboxylate
Supplier:
Apollo Scientific
About this item
CAS Number:
182628-12-2
:
Formula:
C₁₃H₁₇NO₂
MDL Number:
MFCD24368281
:
219.28 g/mol
Storage Temperature:
Ambient