2-Amino-7-methoxy-3-methylquinoline hydrochloride
Catalog # APOSOR307523-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:224.687 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1171863-09-4
Formula:C₁₁H₁₂N₂O·ClH
MDL Number:MFCD10703342