Order Entry
Germany
ContactUsLinkComponent
N-Boc-(S)-2-amino-3-methoxy-1-propanol
Irritant
N-Boc-(S)-2-amino-3-methoxy-1-propanol
Catalog # APOSOR307124-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-Boc-(S)-2-amino-3-methoxy-1-propanol
Catalog # APOSOR307124-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.251 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:721927-59-9
Formula:C₉H₁₉NO₄
MDL Number:MFCD20261175