Order Entry
Germany
ContactUsLinkComponent
Irritant
2,8-Diazaspiro[4.5]decan-3-one hydrochloride
Catalog # APOSOR306483-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,8-Diazaspiro[4.5]decan-3-one hydrochloride
Catalog # APOSOR306483-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :190.671 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:945892-88-6
Formula:C₈H₁₄N₂O·ClH
MDL Number:MFCD11520496