tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate
Catalog # APOSOR305478-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:235.279 g/mol
Storage Temperature:Ambient
CAS Number:957204-30-7
Formula:C₁₃H₁₇NO₃
MDL Number:MFCD11045243