cis-3-Methylcyclobutan-1-amine hydrochloride
Catalog # APOSOR304367-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:121.608 g/mol
Storage Temperature:Ambient
CAS Number:1093951-55-3
Formula:C₅H₁₁N·ClH
MDL Number:MFCD12925785