Order Entry
Germany
ContactUsLinkComponent
cis-3-(Propan-2-yloxy)cyclobutan-1-amine hydrochloride
cis-3-(Propan-2-yloxy)cyclobutan-1-amine hydrochloride
Catalog # APOSOR304342-1G
Supplier:  Apollo Scientific
CAS Number:  
cis-3-(Propan-2-yloxy)cyclobutan-1-amine hydrochloride
Catalog # APOSOR304342-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :165.661 g/mol
Storage Temperature:Ambient
CAS Number:1909294-44-5
Formula:C₇H₁₅NO·ClH
MDL Number:MFCD28892829