Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diethyl 2-(4-methoxyphenyl)malonate
Diethyl 2-(4-methoxyphenyl)malonate
Diethyl 2-(4-methoxyphenyl)malonate
Irritant
Diethyl 2-(4-methoxyphenyl)malonate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Diethyl 2-(4-methoxyphenyl)malonate
Supplier:
Apollo Scientific
About this item
CAS Number:
23197-67-3
:
Formula:
C₁₄H₁₈O₅
MDL Number:
MFCD04038965
:
266.29 g/mol
Boiling Pt:
145…148 °C (1 mm Hg)
:
GHS07
Storage Temperature:
Ambient