Order Entry
Germany
ContactUsLinkComponent
 
Ethyl 6-(aminomethyl)nicotinate hydrochloride 95+%
Catalog # APOSOR301304-250MG
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 6-(aminomethyl)nicotinate hydrochloride 95+%
Catalog # APOSOR301304-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :216.665 g/mol
Storage Temperature:Ambient
CAS Number:1189434-55-6
Formula:C₉H₁₂N₂O₂·ClH
MDL Number:MFCD09788628