Methyl 2-methyl-6-nitrobenzoate
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Methyl 6-nitro-o-toluate, 2-(Methoxycarbonyl)-3-nitrotoluene, 2-(Methoxycarbonyl)-3-methylnitrobenzene
- CAS Number:61940-22-5
- Formula:C₉H₉NO₄
- MDL Number:MFCD11042453
- Molecular Weight:195.172 g/mol
- Melting Pt:50 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: Methyl 6-nitro-o-toluate, 2-(Methoxycarbonyl)-3-nitrotoluene, 2-(Methoxycarbonyl)-3-methylnitrobenzene
- CAS Number:61940-22-5
- Formula:C₉H₉NO₄
- MDL Number:MFCD11042453
- Molecular Weight:195.172 g/mol
- Melting Pt:50 °C
- Storage Temperature:Ambient



