
Methyl 5-chloro-2-methyl-1H-indole-3-carboxylate
Catalog # APOSOR300209-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:223.656 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:762288-06-2
Formula:C₁₁H₁₀ClNO₂
MDL Number:MFCD23159660