N-Methylbicyclo[1.1.1]pentan-1-amine hydrochloride
Catalog # APOSOR300175-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:133.619 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1882089-75-9
:1886967-09-4
Formula:C₆H₁₁N·ClH
MDL Number:MFCD31417451