Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Chloroisoquinoline-6-carboxaldehyde
1-Chloroisoquinoline-6-carboxaldehyde
1-Chloroisoquinoline-6-carboxaldehyde
APOSOR300074-1G
:
Apollo Scientific
:
1-Chloroisoquinoline-6-carboxaldehyde
APOSOR300074-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
191.614 g/mol
:
GHS07
:
Refrigerator
:
1211528-19-6
:
C₁₀H₆ClNO
:
MFCD15526625