Ethyl 5-(4-aminophenyl)-2-methyl-3-furoate
Catalog # APOSOR29475-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:245.274 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:175276-70-7
Formula:C₁₄H₁₅NO₃
MDL Number:MFCD00276981