Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3,6-dichloropyridine-2-carboxylate
Methyl 3,6-dichloropyridine-2-carboxylate 97%
Methyl 3,6-dichloropyridine-2-carboxylate 97%
Irritant
Methyl 3,6-dichloropyridine-2-carboxylate 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 3,6-dichloropyridine-2-carboxylate 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1532-24-7
:
Formula:
C₇H₅Cl₂NO₂
MDL Number:
MFCD00186642
:
206.026 g/mol
:
GHS07
Storage Temperature:
Ambient