Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol
APOSOR27530-25G
:
Apollo Scientific
:
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol
APOSOR27530-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
226.398 g/mol
:
Ambient
:
163119-16-2
:
C₁₅H₃₀O
:
MFCD00278493