
[4-({{(9H-Fluoren-9-yl)methoxy]carbonyl}amino)tetrahydro-2H-pyran-4-yl]acetic acid
Catalog # APOSOR2670-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:381.422 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:946716-25-2
Formula:C₂₂H₂₃NO₅
MDL Number:MFCD09264206