1-(3-Aminoprop-1-yl)-5-(tert-butyl)-2-methyl-1H-pyrrole-3-carboxylic acid, 1-BOC protected
Catalog # APOSOR2441-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:338.442 g/mol
Melting Pt:154…156 °C
:GHS07
Storage Temperature:Ambient
CAS Number:306936-18-5
Formula:C₁₈H₃₀N₂O₄
MDL Number:MFCD01567234