Order Entry
Germany
ContactUsLinkComponent
Methyl 4-methylthiophene-2-carboxylate
Irritant
Methyl 4-methylthiophene-2-carboxylate
Catalog # APOSOR24339-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 4-methylthiophene-2-carboxylate
Catalog # APOSOR24339-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :156.202 g/mol
Boiling Pt:62…68 °C (0.2 mm Hg)
 :GHS07
Storage Temperature:Refrigerator
CAS Number:28686-90-0
Formula:C₇H₈O₂S
MDL Number:MFCD01763641