Order Entry
Germany
ContactUsLinkComponent
Methyl piperazine-2-carboxylate, N1-CBZ protected 97%
Methyl piperazine-2-carboxylate, N1-CBZ protected 97%
Catalog # APOSOR2391-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl piperazine-2-carboxylate, N1-CBZ protected 97%
Catalog # APOSOR2391-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :278.304 g/mol
Storage Temperature:Ambient
CAS Number:126937-43-7
Formula:C₁₄H₁₈N₂O₄
MDL Number:MFCD04115344