Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-5-nitroanisole
3-Chloro-5-nitroanisole 95%
3-Chloro-5-nitroanisole 95%
:
Apollo Scientific
3-Chloro-5-nitroanisole 95%
:
Apollo Scientific
:
55910-07-1
:
:
C₇H₆ClNO₃
:
MFCD11848441
:
187.58 g/mol
:
GHS07
:
Ambient