Order Entry
Germany
ContactUsLinkComponent
3-[2-(Morpholin-4-yl)ethoxy]benzaldehyde hydrochloride
Corrosive
Irritant
3-[2-(Morpholin-4-yl)ethoxy]benzaldehyde hydrochloride
Catalog # APOSOR18570-250MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
3-[2-(Morpholin-4-yl)ethoxy]benzaldehyde hydrochloride
Catalog # APOSOR18570-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :271.74 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
 :GHS05
 :GHS07
CAS Number:1609402-82-5
Formula:C₁₃H₁₇NO₃·ClH
MDL Number:MFCD13186601