tert-Butyl 4-(3-amino-4-nitrophenyl)piperazine-1-carboxylate
Catalog # APOSOR183523-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:322.36 g/mol
Storage Temperature:Ambient
CAS Number:193902-98-6
Formula:C₁₅H₂₂N₄O₄
MDL Number:MFCD12022589