Order Entry
Germany
ContactUsLinkComponent
Ethyl 6-tert-butyl-4-hydroxyquinoline-3-carboxylate
Ethyl 6-tert-butyl-4-hydroxyquinoline-3-carboxylate
Catalog # APOSOR183476-250MG
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 6-tert-butyl-4-hydroxyquinoline-3-carboxylate
Catalog # APOSOR183476-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :273.327 g/mol
Storage Temperature:Ambient
CAS Number:228728-32-3
Formula:C₁₆H₁₉NO₃
MDL Number:MFCD24843129