Order Entry
Germany
ContactUsLinkComponent
3-Bromo-5-nitro-1H-indole, N-BOC protected 98%
3-Bromo-5-nitro-1H-indole, N-BOC protected 98%
Catalog # APOSOR1727-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-5-nitro-1H-indole, N-BOC protected 98%
Catalog # APOSOR1727-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :341.157 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:914349-31-8
Formula:C₁₃H₁₃BrN₂O₄
MDL Number:MFCD05864776