Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Oxa-7-azaspiro[4.5]decan-2-one hydrochloride
1-Oxa-7-azaspiro[4.5]decan-2-one hydrochloride
1-Oxa-7-azaspiro[4.5]decan-2-one hydrochloride
1-Oxa-7-azaspiro[4.5]decan-2-one hydrochloride
APOSOR16705-250MG
:
Apollo Scientific
:
1-Oxa-7-azaspiro[4.5]decan-2-one hydrochloride
APOSOR16705-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
191.655 g/mol
:
GHS07
:
Ambient
:
1314961-56-2
:
C₈H₁₃NO₂·ClH
:
MFCD18642911