Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Octahydro-4H-isoindol-4-one, N-BOC protected
Octahydro-4H-isoindol-4-one, N-BOC protected 95%
Octahydro-4H-isoindol-4-one, N-BOC protected 95%
Irritant
Octahydro-4H-isoindol-4-one, N-BOC protected 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Octahydro-4H-isoindol-4-one, N-BOC protected 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
879687-92-0
:
Formula:
C₁₃H₂₁NO₃
MDL Number:
MFCD17170012
:
239.311 g/mol
:
GHS07
Storage Temperature:
Ambient