
1-Methylindoline-5-carboxaldehyde 97%
Catalog # APOSOR14889-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:161.2 g/mol
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
CAS Number:60082-02-2
Formula:C₁₀H₁₁NO
MDL Number:MFCD03208513