

1-Methyl-5-[(methylamino)methyl]indoline 90%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:906352-81-6
:
Formula:C₁₁H₁₆N₂
MDL Number:MFCD09817499
:176.258 g/mol
UN:2922
ADR:8+6.1,III
:GHS05
:GHS07
Storage Temperature:Refrigerator