Order Entry
Germany
ContactUsLinkComponent
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carbonyl chloride
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carbonyl chloride
Catalog # APOSOR1281-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carbonyl chloride
Catalog # APOSOR1281-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Boiling Pt:117 °C (15 mm Hg)
Storage Temperature:Ambient
UN:3265
ADR:8,III
 :200.665 g/mol
 :GHS05
 :GHS07
CAS Number:160842-62-6
Formula:C₉H₁₃ClN₂O
MDL Number:MFCD00084916