Order Entry
Germany
ContactUsLinkComponent
 
Irritant
1-Benzyl 2-methyl (2S,4S)-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride
Catalog # APOSOR12769-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Benzyl 2-methyl (2S,4S)-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride
Catalog # APOSOR12769-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :314.765 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1212395-57-7
Formula:C₁₄H₁₈N₂O₄·ClH
MDL Number:MFCD06804628