
Biphenyl-2-carboxaldehyde
Catalog # APOSOR12525-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.218 g/mol
Boiling Pt:91…92 °C (0.15 mm Hg)
Density:1.13 g/cm³ (25 °C)
Flash Pt:102 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:1203-68-5
Formula:C₁₃H₁₀O
MDL Number:MFCD01740431