
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1135-24-6
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00004400
:194.18 g/mol
Melting Pt:169…173 °C
EINECS:214-490-0
:GHS07
Storage Temperature:Ambient