Ethyl 2-(3-nitrophenyl)pyridine-3-carboxylate
Catalog # APOSOR111536-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:272.256 g/mol
Storage Temperature:Ambient
CAS Number:2244083-82-5
Formula:C₁₄H₁₂N₂O₄
MDL Number:MFCD31629941