4-Amino-N-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)benzenesulphonamide
Catalog # APOSOR111058-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:322.383 g/mol
Storage Temperature:Ambient
CAS Number:1858251-36-1
Formula:C₁₄H₁₈N₄O₃S
MDL Number:MFCD28148230