tert-Butyl (5-nitro-1H-indol-1-yl)acetate
Catalog # APOSOR110901-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.288 g/mol
Storage Temperature:Ambient
CAS Number:227302-34-3
Formula:C₁₄H₁₆N₂O₄
MDL Number:MFCD27995627