Order Entry
Germany
ContactUsLinkComponent
Methyl (1-oxo-1,3-dihydro-2H-isoindol-2-yl)acetate
Methyl (1-oxo-1,3-dihydro-2H-isoindol-2-yl)acetate
Catalog # APOSOR110871-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl (1-oxo-1,3-dihydro-2H-isoindol-2-yl)acetate
Catalog # APOSOR110871-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.21 g/mol
Storage Temperature:Ambient
CAS Number:96017-03-7
Formula:C₁₁H₁₁NO₃
MDL Number:MFCD04124610