Order Entry
Germany
ContactUsLinkComponent
3,4-Dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylic acid
Irritant
3,4-Dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylic acid
Catalog # APOSOR110753-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,4-Dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylic acid
Catalog # APOSOR110753-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
Melting Pt:207…217 °C
 :207.183 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:141762-02-9
Formula:C₁₀H₉NO₄
MDL Number:MFCD12756239