tert-Butyl 2-[(3-bromobenzoyl)amino]ethylcarbamate
Catalog # APOSOR110646-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:343.216 g/mol
Storage Temperature:Ambient
CAS Number:1427460-40-9
Formula:C₁₄H₁₉BrN₂O₃
MDL Number:MFCD18804161