tert-Butyl 4-[2-(2-formylphenoxy)ethyl]piperazine-1-carboxylate
Catalog # APOSOR110582-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:334.41 g/mol
Storage Temperature:Ambient
CAS Number:1227954-66-6
Formula:C₁₈H₂₆N₂O₄
MDL Number:MFCD11973599