0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Methyl 1,3-benzoxazole-6-carboxylate
Methyl 1,3-benzoxazole-6-carboxylate 95%
Methyl 1,3-benzoxazole-6-carboxylate 95%
APOSOR110295-1G
:
Apollo Scientific
:
Methyl 1,3-benzoxazole-6-carboxylate 95%
APOSOR110295-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
177.157 g/mol
:
Ambient
:
1305711-40-3
:
C₉H₇NO₃
:
MFCD14635902